Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aldolase A Antibody (3F9), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242620F
Description
Aldolase A Monoclonal specifically detects Aldolase A in Human, Mouse samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Aldolase A | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
ALDOA | |
Recombinant human ALDOA (1-364aa) purified from E. coli | |
100 μL | |
Cancer, Cell Cycle and Replication | |
226 | |
Store at 4C in the dark. | |
IgG2a κ |
Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
3F9 | |
Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
ALDA, aldolase A, fructose-bisphosphate, EC 4.1.2.13, fructose-1,6-bisphosphate triosephosphate-lyase, fructose-bisphosphate aldolase A, GSD12, Lung cancer antigen NY-LU-1, MGC10942, MGC17716, MGC17767, Muscle-type aldolase | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction