Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aldolase A/B/C Monoclonal Antibody (E9)

Catalog No. PIMA547377
Change view
Click to view available options
Quantity:
100 μL
500 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547377 500 μL
PIMA547476 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA547377 Supplier Invitrogen™ Supplier No. MA547377
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Aldolase A/B/C Monoclonal Antibody for Western Blot, ICC/IF, IHC

Aldolases are important glycolytic cytosolic enzymes which catalyse the reversible conversion of fructose-1,6-bisphosphate to glyceraldehyde 3-phosphate. There are three aldolase isozymes coded by three distinct genes in mammals, namely aldolases A, B, and C. Aldolase A is heavily expressed in muscle enzyme and aldolase B is heavily expressed in liver and is regarded as a liver specific enzyme. In the adult, aldolase C is the brain-specific isozyme expressed in astrocytes and a few classes of neurons, notably Purkinje cells. Appropriate antibodies to aldolase C are therefore useful to identify astrocytes in cell culture and sectioned material, and the enzyme may be over expressed in some forms of cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aldolase A/B/C
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone E9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 5mM sodium azide
Gene ALDOC
Gene Accession No. P00884, P04075, P05062, P05063, P05064, P05065, P09117, P09972, Q91Y97
Gene Alias ALDA; ALDB; ALDC
Gene Symbols ALDOA, ALDOB, ALDOC
Host Species Mouse
Immunogen Recombinant human full-length aldolase C.
Purification Method Protein A/G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11674, 11676, 226, 229, 230, 230163, 24189, 24190, 24191
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.