Learn More
Invitrogen™ Aldolase A/B/C Monoclonal Antibody (E9)

Description
Aldolase A/B/C Monoclonal Antibody for Western Blot, ICC/IF, IHC
Specifications
Specifications
| Antigen | Aldolase A/B/C |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | E9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 5mM sodium azide |
| Gene | ALDOC |
| Gene Accession No. | P00884, P04075, P05062, P05063, P05064, P05065, P09117, P09972, Q91Y97 |
| Gene Alias | ALDA; ALDB; ALDC |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.