Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aldolase A Recombinant Rabbit Monoclonal Antibody (7D8)
GREENER_CHOICE

Catalog No. FEMA549889
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA549889 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA549889 Supplier Invitrogen™ Supplier No. MA549889
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Aldolase A Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Aldolase encodes a member of the class I fructose-bisphosphate aldolase protein family. The encoded protein is a glycolytic enzyme that catalyzes the reversible conversion of fructose-1,6-bisphosphate to glyceraldehyde 3-phosphate and dihydroxyacetone phosphate. Three aldolase isozymes (A, B, and C), encoded by three different genes, are differentially expressed during development. Mutations in this gene have been associated with Glycogen Storage Disease XII, an autosomal recessive disorder associated with hemolytic anemia. Disruption of this gene also plays a role in the progression of multiple types of cancers. Related pseudogenes have been identified on chromosomes 3 and 10.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aldolase A
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 7D8
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ALDOA
Gene Accession No. P04075, P05064, P05065
Gene Alias ALDA; Aldo1; Aldo-1; ALDOA; Aldolase 1; aldolase 1, A isoform; aldolase A; aldolase A, fructose-bisphosphate; aldolase, fructose-bisphosphate A; epididymis secretory sperm binding protein Li 87p; fructose-1,6-bisphosphate triosephosphate-lyase; fructose-bisphosphate aldolase A; GSD12; HEL-S-87p; Lung cancer antigen NY-LU-1; muscle-type aldolase; RNALDOG5; unnamed protein product
Gene Symbols ALDOA
Host Species Rabbit
Immunogen A synthesized peptide derived from human Aldolase.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11674, 226, 24189
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.