Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aldolase B Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297910PCP
Description
Aldolase B Polyclonal antibody specifically detects Aldolase B in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Aldolase B | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
ALDO2, aldolase B, fructose-bisphosphate, EC 4.1.2.13, fructose-bisphosphatase, fructose-bisphosphate aldolase B | |
Produced in rabbits immunized with E. coli-derived Human Aldolase B fragment. | |
0.1 mL | |
Cancer | |
229 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction