Learn More
Invitrogen™ Aldolase B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA592047
Description
Immunogen sequence: MAHRFPALTQ EQKKELSEIA QSIVANGKGI LAADESVGTM GNRLQRIKVE NTEENRRQFR EILFSVDSSI NQSIGGVILF HETLYQKDSQ GKLFRNILKE KGIVVGIKLD QGGAPLAGTN KETTIQGLDG LSERCAQYKK DGVDFGKWRA VLRIADQCPS SLAIQENANA LARYASICQQ NGLVPIVEPE VIPDGDHDLE; Positive Samples: LO2, K-562, MCF7, Mouse liver, Mouse kidney, Mouse skeletal muscle, Rat liver, Rat kidney, Mouse heart; Cellular Location: Cytoplasm, centriolar satellite, centrosome, cytoskeleton, microtubule organizing center.
Fructose-1,6-bisphosphate aldolase is a tetrameric glycolytic enzyme that catalyzes the reversible conversion of fructose-1,6-bisphosphate to glyceraldehyde 3-phosphate and dihydroxyacetone phosphate. Vertebrates have 3 aldolase isozymes which are distinguished by their electrophoretic and catalytic properties. Differences indicate that aldolases A, B, and C are distinct proteins, the products of a family of related 'housekeeping' genes exhibiting developmentally regulated expression of the different isozymes. The developing embryo produces aldolase A, which is produced in even greater amounts in adult muscle where it can be as much as 5% of total cellular protein. In adult liver, kidney and intestine, aldolase A expression is repressed and aldolase B is produced. In brain and other nervous tissue, aldolase A and C are expressed about equally. There is a high degree of homology between aldolase A and C. Defects in ALDOB cause hereditary fructose intolerance.
Specifications
Aldolase B | |
Polyclonal | |
Unconjugated | |
Aldob | |
ALDB; Aldo2; Aldo-2; ALDOB; Aldolase 2; aldolase 2, B isoform; Aldolase B fructose-biphosphate; Aldolase B, fructose-biphosphate; aldolase B, fructose-bisphosphatase; aldolase B, fructose-bisphosphate; aldolase, fructose-bisphosphate B; BC016435; Fructose-bisphosphate aldolase B; LIV10; Liver-type aldolase | |
Rabbit | |
Affinity Chromatography | |
RUO | |
229, 230163, 24190 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.59 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P00884, P05062, Q91Y97 | |
Aldob | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human ALDOB (NP_0000262). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.