Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aldolase B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534659
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534659 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534659 Supplier Invitrogen™ Supplier No. PA534659
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, HeLa, HepG2, mouse brain, rat brain, Neuro2A, GL261, C8D30, PC-12. Predicted reactivity: Mouse (96%), Rat (97%), Rabbit (98%), Chicken (84%), Sheep (90%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Fructose-1,6-bisphosphate aldolase is a tetrameric glycolytic enzyme that catalyzes the reversible conversion of fructose-1,6-bisphosphate to glyceraldehyde 3-phosphate and dihydroxyacetone phosphate. Vertebrates have 3 aldolase isozymes which are distinguished by their electrophoretic and catalytic properties. Differences indicate that aldolases A, B, and C are distinct proteins, the products of a family of related 'housekeeping' genes exhibiting developmentally regulated expression of the different isozymes. The developing embryo produces aldolase A, which is produced in even greater amounts in adult muscle where it can be as much as 5% of total cellular protein. In adult liver, kidney and intestine, aldolase A expression is repressed and aldolase B is produced. In brain and other nervous tissue, aldolase A and C are expressed about equally. There is a high degree of homology between aldolase A and C. Defects in ALDOB cause hereditary fructose intolerance.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aldolase B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.09 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene Aldob
Gene Accession No. P00884, P05062, Q91Y97
Gene Alias ALDB; Aldo2; Aldo-2; ALDOB; Aldolase 2; aldolase 2, B isoform; Aldolase B fructose-biphosphate; Aldolase B, fructose-biphosphate; aldolase B, fructose-bisphosphatase; aldolase B, fructose-bisphosphate; aldolase, fructose-bisphosphate B; BC016435; Fructose-bisphosphate aldolase B; LIV10; Liver-type aldolase
Gene Symbols Aldob
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Aldolase B. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 229, 230163, 24190
Target Species Human, Mouse, Reptile, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.