Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aldolase C Antibody (1A1), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP225144PE
Description
Aldolase C Monoclonal specifically detects Aldolase C in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
Aldolase C | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
ALDOC | |
The C-terminal 23 amino acids of human Aldolase C (KYEGSGEDGGAAAQSLYIANHAY). [UniProt# P09972] | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
230 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
1A1 | |
Flow Cytometry | |
ALDC, aldolase 3, aldolase C, fructose-bisphosphate, Brain-type aldolase, EC 4.1.2.13, fructoaldolase C, fructose-1,6-biphosphate triosephosphate lyase, fructose-bisphosphate aldolase C | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction