Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aldolase C Mouse anti-Human, PerCP, Clone: 200000000000, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259408PCP
This item is not returnable.
View return policy
Description
Aldolase C Monoclonal antibody specifically detects Aldolase C in Human samples. It is validated for Flow CytometrySpecifications
Aldolase C | |
Monoclonal | |
PerCP | |
ALDC, aldolase 3, aldolase C, fructose-bisphosphate, Brain-type aldolase, EC 4.1.2.13, fructoaldolase C, fructose-1,6-biphosphate triosephosphate lyase, fructose-bisphosphate aldolase C | |
Recombinant human Aldolase C (1-364aa) purified from E. coli | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
230 | |
Store at 4°C in the dark. | |
IgG1 κ |
Flow Cytometry | |
200000000000 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction