Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aldolase C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114842
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114842 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114842 Supplier Invitrogen™ Supplier No. PA5114842
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ALDOC.

This gene encodes a member of the class I fructose-biphosphate aldolase gene family. Expressed specifically in the hippocampus and Purkinje cells of the brain, the encoded protein is a glycolytic enzyme that catalyzes the reversible aldol cleavage of fructose-1,6-biphosphate and fructose 1-phosphate to dihydroxyacetone phosphate and either glyceraldehyde-3-phosphate or glyceraldehyde, respectively. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aldolase C
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene ALDOC
Gene Accession No. P05063, P09117, P09972
Gene Alias AI847350; ALDC; ALDCAA; Aldo3; ALDOC; Aldolase 3; aldolase C, fructose-biphosphate; aldolase C, fructose-bisphosphate; aldolase, fructose-bisphosphate C; AU040929; Brain-type aldolase; EC 4.1.2.13; F16dip7; fructoaldolase C; fructose-1,6-biphosphate triosephosphate lyase; fructose-bisphosphate aldolase C; RATALDCAA; Scrapie-responsive protein 2; Scrg2; Zebrin II
Gene Symbols ALDOC
Host Species Rabbit
Immunogen A synthesized peptide derived from human ALDOC(Accession P09972), corresponding to amino acid residues R294-E344.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11676, 230, 24191
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.