Learn More
Aldolase C Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA551883
Description
Immunogen sequence: APFDKKCGRE NWTVAFAPDG SYFAWSQGHR TVKLVPWSQC LQNFLLHGTK NVTNSSSLRL PRQNSDGGQK NKPREHIIDC GDIVWSLAFG SSVPEKQSRC VNIEWHRFRF GQDQLLLATG LNNGRIKIWD V Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 9
This gene encodes a member of the class I fructose-biphosphate aldolase gene family. Expressed specifically in the hippocampus and Purkinje cells of the brain, the encoded protein is a glycolytic enzyme that catalyzes the reversible aldol cleavage of fructose-1,6-biphosphate and fructose 1-phosphate to dihydroxyacetone phosphate and either glyceraldehyde-3-phosphate or glyceraldehyde, respectively. [provided by RefSeq, Jul 2008]
Specifications
Aldolase C | |
Polyclonal | |
Unconjugated | |
ALDOC | |
ALDC, aldolase 3, brain-type aldolase, fructoaldolase C, fructose-1,6-biphosphate triosephosphate lyase, fructose-bisphosphate aldolase C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11676, 230, 24191 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P09972, P05063, P09117 | |
ALDOC | |
Recombit protein corresponding to Human Aldolase C | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Research Use Only