Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aldolase C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | Aldolase C |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP191323
|
Novus Biologicals
NBP191323 |
100 μL |
Each of 1 for $482.50
|
|
|||||
Description
Aldolase C Polyclonal specifically detects Aldolase C in Human samples. It is validated for Western Blot.Specifications
Aldolase C | |
Polyclonal | |
Rabbit | |
NP_005156 | |
230 | |
Synthetic peptide directed towards the C terminal of human ALDOC. Peptide sequence CPLPRPWALTFSYGRALQASALNAWRGQRDNAGAATEEFIKRAEVNGLAA. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
ALDC, aldolase 3, aldolase C, fructose-bisphosphate, Brain-type aldolase, EC 4.1.2.13, fructoaldolase C, fructose-1,6-biphosphate triosephosphate lyase, fructose-bisphosphate aldolase C | |
ALDOC | |
IgG | |
39 kDa |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title