Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Innovating Science™ Freeze Dried Bacteria Cultures
Shop All Aldon Products

Click to view available options
Product Type:
Bacillus subtilis
E. coli
Micrococcus luteus
Serratia marcescens
Staphylococcus epidermidis
Description
Includes
One tube of rehydrating broth and one tube agar slant, sterile swab and instructions.
- Lyophilized pellet form in vial
- Available for five different types of bacteria
Specifications
Specifications
Color | White |
Grade | 7 to 14 |
Age | 13 to 20 |
For Use With (Application) | For use in classroom for educational purposes |
Includes | One tube of rehydrating broth and one tube agar slant, sterile swab and instructions |
Quantity | 1 |
Class Size | 1 Class |
Product Type | Staphylococcus epidermidis |
Description | Format: Lyophilized pellet |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction