Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Green Fuel Cell: Energy From Yeast Kit
SCI_ED

Catalog No. s06971
Click to view available options
:
Green Fuel Cell: Energy from Yeast Kit; For 15 groups
WARNING: This product can expose you to chemicals including Phthalates, which is known to the State of California to cause and reproductive harm.
WARNING: This product can expose you to chemicals including Phthalates, which is known to the State of California to cause and reproductive harm.

Includes

  • Yeast instant dry (10g)
  • Dextrose R/G anhydrous (D-Glucose) (25g)
  • Phosphate buffer pH 7.0 (7.5g)
  • Potassium ferricyanide 1.0M (5mL)
  • Methylene blue chloride 1% Aq (5mL)
  • 15mL Bottles (30)
  • Connecting wire, red 12 in. with alligator clips (15)
  • Connecting wire, black 12 in. with alligator clips (15)
  • Carbon electrodes (30)
  • 0.75 x 4 in. strips, filter paper 8 x 11 in.
  • Teacher's manual and student study guide copymasters

Learn how biological organisms create energy.

Electron transfer is part of the metabolic process in living organisms.

  • Set up a simple cell using Yeast
  • Use special dye that enter into cells to collect electron and determine if the haevested electrons are capable of producing current in the cell and if so how much current

Order Info

For 15 groups

Specifications

Certifications/Compliance 49 CFR 173.4
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Learn how biological organisms create energy
Includes 10g Yeast instant dry, 25g Dextrose R/G Anhydrous (D-Glucose), 7.5g Phosphate Buffer pH 7, 5mL Potassium Ferricyanide 1M, 5mL Methylene Blue Chloride 1%, 30 15mL bottles, 15 bottle caps, 15 connecting wire, red 12 in. with alligator clips, 15 connecting wire, black 12 in. with alligator clips, 30 Carbon electrodes, 20 0.25 X 4 in. strips, 8 X 11 in. filter paper, Teacher's manual and student study guide copymasters
Quantity 1
Dimensions (L x W x H) 11.125 x 8.75 x 4 in.
Class Size 15 Lab Groups
Product Type Green Fuel Cell Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.