Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aldosterone Antibody (BGN/10/42110), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10062313G
Description
Aldosterone Monoclonal specifically detects Aldosterone in All Species samples. It is validated for ELISA.Specifications
Aldosterone | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Aldosterone (3-CMO) conjugated to Bovine Serum Albumin. | |
0.1 mL | |
Primary | |
All species | |
Purified |
ELISA | |
BGN/10/42110 | |
ELISA | |
Mouse | |
Protein A or G purified | |
RUO | |
This product is specific for aldosterone and exhibits minimal reactivity with androstenedione, corticosterone, DHEA and desoxycortisol. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction