Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aldosterone Antibody (BGN/10/4213), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10062316AF532
Description
Aldosterone Monoclonal specifically detects Aldosterone in All Species samples. It is validated for ELISA.Specifications
Aldosterone | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
Aldosterone conjugated to Bovine Serum Albumin. | |
0.1 mL | |
Aldosterone. This antibody shows less than 0.01% cross-reactivity with Androstenedione, Corticosterone, DHEA and Deoxcorticosterone. | |
Store at 4°C in the dark. |
ELISA | |
BGN/10/4213 | |
ELISA | |
Mouse | |
Protein A purified | |
Primary | |
All species | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction