Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aldosterone Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$170.97 - $492.89
Specifications
| Antigen | Aldosterone |
|---|---|
| Concentration | 5.0 mg/mL |
| Dilution | ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017626
![]() |
Novus Biologicals
NB100646580.1ML |
0.1 mL |
Each for $170.97
|
|
|||||
NB10064658
![]() |
Novus Biologicals
NB10064658 |
0.5 mL |
Each for $492.89
|
|
|||||
Description
Aldosterone Polyclonal specifically detects Aldosterone in Multi-Species samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
| Aldosterone | |
| ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Multi-species | |
| Aldosterone-3-CMO-BSA | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 5.0 mg/mL | |
| ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS (pH 7.2) with 0.09% Sodium Azide | |
| IgG | |
| Protein G purified | |
| Aldosterone 100% Androstenedione <1% Corticosterone <1% DHEA <1% Desoxycorticosterone <1% |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title