Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aldosterone Monoclonal Antibody (BGN/10/42110)
GREENER_CHOICE

Catalog No. MA182846
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA182846 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA182846 Supplier Invitrogen™ Supplier No. MA182846
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody shows minimal cross-reactivity with androstenedione, corticosterone, DHEA, and desoxycortisol. Mouse anti aldosterone antibody, clone 1-10.1 recognizes aldosterone, a mineralocorticocoid hormone essential for sodium and potassium homeostasis. Mouse anti aldosterone antibody, clone 1-10.1 exhibits minimal reactivity with androstenedione, corticosterone, DHEA and desoxycortisol.

Aldosterone is a hormone produced by the adrenal glands that helps regulate blood pressure through increasing both the release of potassium and the uptake of water and sodium by the kidneys. It is essential for sodium conservation in the kidney, salivary glands, sweat glands and colon. It plays a central role in the regulation of the plasma sodium (Na+), the extracellular potassium (K+) and arterial blood pressure. It does so mainly by acting on the mineralocorticoid receptors in the distal tubules and collecting ducts of the nephron. It influences the reabsorption of sodium and excretion of potassium (from and into the tubular fluids, respectively) of the kidney, thereby indirectly influencing water retention or loss, blood pressure and blood volume. When dysregulated, aldosterone is pathogenic and contributes to the development and progression of cardiovascular and renal disease. Aldosterone has exactly the opposite function of the atrial natriuretic hormone secreted by the heart.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aldosterone
Applications ELISA
Classification Monoclonal
Clone BGN/10/42110
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Host Species Mouse
Immunogen Aldosterone (3-CMO) conjugated to BSA.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.