Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alendronic Acid 98.0+%, TCI America™
SDP

Catalog No. A212025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A212025G 25 g
A21205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A212025G Supplier TCI America Supplier No. A212025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 66376-36-1
Molecular Formula C4H13NO7P2
Molecular Weight (g/mol) 249.096
MDL Number MFCD00868112
InChI Key OGSPWJRAVKPPFI-UHFFFAOYSA-N
Synonym 4-Amino-1-hydroxybutane-1,1-diphosphonic Acid
PubChem CID 2088
ChEBI CHEBI:2567
IUPAC Name (4-amino-1-hydroxy-1-phosphonobutyl)phosphonic acid
SMILES C(CC(O)(P(=O)(O)O)P(=O)(O)O)CN

Specifications

Melting Point 235°C
Color White-Yellow
Quantity 25 g
UN Number 1759
Formula Weight 249.10
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Alendronic Acid
RTECSNumber SZ6521833
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.