Learn More
Abbott™ Acceava™ Mono Cassette Test
Catalog No. 23046670 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Quantity:
25 Cassettes
Catalog No. Quantity
23-046-670 25 Cassettes
1 options
This item is not returnable. View return policy
Catalog No. 23-046-670 Supplier Abbott™ Supplier No. 92407
This item is not returnable. View return policy
Includes
  • 25 Acceava Mono Cassette test devices
  • 25 (10μL) (black line) Sample transfer pipettes for use with serum/plasma
  • 25 (25μL) (red line) Sample transfer pipettes for use with whole blood
  • 1 Developer solution
  • 1 Negative control
  • 1 Positive control
  • 1 Package insert
  • 1 Procedure card
Requires
  • Centrifuge capable of separation of blood cells from plasma
  • Lancet
Provides superior performance for the diagnosis of mononucleosis. One-step antibody test for IM that uses direct solid-phase immunoassay technology to detect IM heterophile antibodies in human serum, plasma, or whole blood.
  • Rapid, flexible procedure reduces office staff hands-on time and follow-up activities
  • One product available for both CLIA-Moderate and Waived labs
  • Walk-away flexibility
  • Positive results in 8 minutes
  • Decrease unnecessary use of antibiotics
  • Decrease need for ancillary testing
Specifications
Clia Complexity Waived, Moderate
Format Cassette
Type Mononucleosis Cassette Test
Includes 25 Mono cassette test devices, 25 x 10μL (black line) sample transfer pipettes (Serum/Plasma), 25 x 25μL (red line) sample transfer pipettes (whole Blood), 1 developer solution, 1 negative control, 1 positive control, 1
Quantity 25 Cassettes