Learn More
Abbott™ AlcoMate™ Premium 7000 Breath Alcohol Tester Kit
Catalog No. 22911077
Click to view available options
:
AlcoMate Premium 7000 Breath Alcohol Tester
This item is not returnable. View return policy
This item is not returnable. View return policy
Includes
Breathalyzer, User Manual, 2 x AA Batteries, 55 x mouthpieces, 1 pre-installed Sensor Module (PRISM™), 1 additional Sensor Module, hard plastic carrying case, car adapter, handstrap
DOT approved tester with one button operation that requires no calibration service to use
Certifications
ISO 9002 Certified, Meets U.S. Coast Guard Requirements
Specifications
Type AlcoMate™ Premium 7000 Breath Alcohol Tester
Includes Breathalyzer, User Manual, 2 x AA Batteries, 55 x mouthpieces, 1 pre-installed Sensor Module (PRISM™), 1 additional Sensor Module, hard plastic carrying case, car adapter, handstrap
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.