Learn More
C. DIFF QUIK CHEK COMPLETE™
Catalog No. p-3797018
Click to view available options
Quantity:
25 Tests
50 Tests
This item is not returnable. View return policy
Catalog No. 23-900-552
Supplier: Techlab Inc 30525C
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
The C. DIFF QUIK CHEK COMPLETE™ test is a rapid membrane enzyme immunoassay for the simultaneous detection of Clostridioides difficile glutamate dehydrogenase antigen and toxins A and B in a single reaction well. Developed and Manufactured by TECHLAB, Inc.
  • Results in approximately 30 minutes
  • Easy to intrepret
  • Highly sensitive and specific
Specifications
Detectable Analytes GDH Antigen, Toxins A and B
Type Clostridium difficile QUIK CHEK COMPLETE™
Includes 25 Membrane device pouches, diluent, wash buffer, conjugate, substrate, positive control, disposable plastic transfer pipettes
Quantity 25 Tests
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.