Learn More
Abbott™ Clearview™ hCG Tests
Catalog No. 23046596
Click to view available options
Clia Complexity:
Waived
Waived (Urine), Non-waived (Serum)
Detectable Analytes:
hCG
Quantity:
40 Tests
50 Tests
Type:
25 hCG Combo Cassette
hCG Combo II Cassette
hCG Urine Cassettes
hCG Urine Dipstick
This item is not returnable. View return policy
This item is not returnable. View return policy
Innovative one-step tests provide highly sensitive and specific results for the early detection of the human pregnancy hormone hCG
  • CLIA Complexity: Waived for urine, Non-Waived for serum
  • Variety of easy-to-use, single-step tests
  • Highly sensitive assay with built-in controls ensures accuracy
  • Test results in under five minutes
Specifications
Clia Complexity Waived
Detectable Analytes hCG
Sample Type Urine
Type hCG Urine Cassettes
Quantity 40 Tests
Test Time 5 min. or less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.