Learn More
Abbott™ Clearview™ hCG Tests
Shop All Abbott Rapid Diagnostics Products
Click to view available options
Clia Complexity:
Waived
Waived (Urine), Non-waived (Serum)
Detectable Analytes:
hCG
Quantity:
40 Tests
50 Tests
Type:
25 hCG Combo Cassette
hCG Combo II Cassette
hCG Urine Cassettes
hCG Urine Dipstick
Description
- CLIA Complexity: Waived for urine, Non-Waived for serum
- Variety of easy-to-use, single-step tests
- Highly sensitive assay with built-in controls ensures accuracy
- Test results in under five minutes
Specifications
Specifications
| Clia Complexity | Waived |
| Detectable Analytes | hCG |
| Sample Type | Urine |
| Type | hCG Urine Dipstick |
| Quantity | 50 Tests |
| Test Time | 5 min. or less |