Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C. Difficile Toxin/Antitoxin Kit
Click to view available options
Quantity:
25 Tests
Description
Includes
Lyophilized C. difficile toxin control reagent (1mL) (cytotoxic titer of >10); Lyophilized C. difficile antitoxin (3mL) (prepared in goats)
Clostridioides difficile produces two toxins, A and B. Toxin B, often referred to as the cytotoxin, is the toxin detected by the tissue culture assay.
- CPT Code: 87230
- Sensitivity: 99.2%
- Specificity: 100%
- Highly sensitive
- Lyophilized reagents
- Minimizes repeated testing of specimens containing high toxin levels
- Intended for use in conjunction with the tissue culture cytotoxicity assay for the confirmation of Clostridioides difficile toxin in patient specimens
- Kit includes a toxin control reagent and a specific C. difficile antitoxin for use in a tissue culture assay
- To be used as an aid in the diagnosis of C. difficile disease; results should be considered in conjunction with the patient history
- For in vitro diagnostic use
Specifications
Specifications
Detectable Analytes | C. difficile toxins A and B |
Type | Clostridium difficile Toxin and Antitoxin Kit |
Includes | Lyophilized C. difficile Toxin Control reagent (1mL) (Cytotoxic Titer of >10), Lyophilized C. difficile Antitoxin (3mL) (Prepared in Goats), Highly sensitive, Lyophilized reagents, Minimizes repeated testing of specimens containing high toxin levels |
Quantity | 25 Tests |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction