Learn More
LACTOFERRIN EZ VUE™
Click to view available options
Quantity:
25 Tests
Description
- Test yields results in 10 minutes
- Identify patients with active inflammatory bowel disease (IBD) and rule out those with active noninflammatory irritable bowel syndrome (IBS)
- Simple procedure
- Accurate measurement of both solid and liquid fecal samples
- Samples can be stored refrigerated or room temperature for up to two weeks prior to testing
Specifications
Specifications
| CPT Code | 83630 |
| Clia Complexity | Moderate |
| Detectable Analytes | Fecal Lactoferrin |
| Type | LACTOFERRIN EZ VUE™ |
| Includes | Diluent, positive control, membrane cassette |
| Quantity | 25 Tests |
| Test Time | 10 min. |
| Form | Liquid |