Learn More
Abbott™ iCassette™ Urine Drug Screen
Click to view available options
Detectable Analytes:
AMP, BAR, BZO, Cocaine (COC), mAMP, MTD, OPI, Oxycodone (OXY), Phencyclidine (PCP), THC (Cannabinoids)
AMP, BAR, BZO, Cocaine (COC), mAMP, MTD, OPI, Phencyclidine (PCP), TCA, THC (Cannabinoids)
AMP, BAR, BZO, Cocaine (COC), mAMP, MTD, OPI, Phencyclidine (PCP), THC (Cannabinoids)
AMP, BAR, BZO, Cocaine (COC), mAMP, OPI, Phencyclidine (PCP), THC (Cannabinoids)
Cocaine (COC), mAMP, OPI, THC (Cannabinoids)
Quantity:
25 Pack
Description
Rapid urine screening test that can be performed without the use of an instrument
- Simple procedure: collect and pipette to test — results in as little as 5 minutes
- Requires only 5mL of specimen
- Tests for up to 10 drugs at once
- Results can be photocopied for record keeping purposes
- 12-18 Month shelf life
- Results are easy to read
- Room temperature storage
Specifications
Specifications
| Detectable Analytes | AMP, BAR, BZO, Cocaine (COC), mAMP, MTD, OPI, Oxycodone (OXY), Phencyclidine (PCP), THC (Cannabinoids) |
| Sample Type | Urine |
| Type | iCassette™ Urine Drug Screen |
| Quantity | 25 Pack |
| Test Time | 5 min. |