Learn More
Abbott™ iScreen™ Dip Drug Cards
Click to view available options
Detectable Analytes:
AMP, BAR, BZO, Cocaine (COC), mAMP, MDMA, MTD, OPI, Oxycodone (OXY), Phencyclidine (PCP), PPX, THC (Cannabinoids)
AMP, BAR, BZO, Cocaine (COC), mAMP, MTD, OPI, Phencyclidine (PCP), TCA, THC (Cannabinoids)
AMP, BAR, BZO, Cocaine (COC), mAMP, MTD, OPI, Phencyclidine (PCP), THC (Cannabinoids)
AMP, BAR, BZO, Cocaine (COC), mAMP, OPI, Phencyclidine (PCP), THC (Cannabinoids)
AMP, Cocaine (COC), mAMP, OPI, THC (Cannabinoids)
Buprenorphine
Cocaine (COC)
Oxycodone (OXY)
Tricyclic Antidepressants
Description
Detect the presence of drugs in minutes
- Simple dip and read procedure
- Room temperature storage
- Laboratory accurate
- 1-12 Drug configurations available
- Results in minutes
- 24 Month shelf life
Specifications
Specifications
| Detectable Analytes | AMP, BAR, BZO, Cocaine (COC), mAMP, MDMA, MTD, OPI, Oxycodone (OXY), Phencyclidine (PCP), PPX, THC (Cannabinoids) |
| Sample Type | Urine |
| Type | iScreen Urine Drug Screen |
| Quantity | 25/Pk. |
| Test Time | Results in minutes |