Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRYPTOSPORIDIUM II™

Catalog No. 23046941
Click to view available options
Quantity:
96 Tests
This item is not returnable. View return policy
This item is not returnable. View return policy

Used to detect Cryptosporidium oocyst antigens in fecal specimens. It is indicated for use as an aid in the diagnosis of patients with diarrhea suspected of Cryptosporidium gastrointestinal infection. Developed and Manufactured by TECHLAB, Inc.

  • Test yields results in less than two hours
  • Assay can be run on the benchtop or automated with Dynex automated systems
  • Break-apart microplate wells minimize reagent use

Specifications

Detectable Analytes Cryptosporidium Antigen
Format Microplate
Sensitivity 97.7%
Includes Conjugate, diluent, positive control, wash solution, stop solution, substrate, microassay plate
Quantity 96 Tests
Test Time <2 hr.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.