Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abbott™ IMPACT RPR™ Needles and Sheaths
Shop All Abbott Rapid Diagnostics Products
Click to view available options
Quantity:
10 Pack
Description
- Classified as a standard test for syphilis by the Centers for Disease Control and Prevention (CDC)
- Non-treponemal test for the serological detection of syphilis
- Assay utilizes charcoal particle technology
Specifications
Specifications
| Type | Needle and Sheath, 20 gauge |
| For Use With (Application) | IMPACT RPR™ Dispense Bottle (61C9) |
| Product Line | IMPACT RPR |
| Quantity | 10 Pack |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction