Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ Alethia™ Mycoplasma Direct DNA Amplification Assay
GSA_VA

Catalog No. 23029012 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-029-012 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-029-012 Supplier Meridian Bioscience™ Supplier No. 480250
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Qualitative in vitro diagnostic test for detection of Mycoplasma pneumoniae, DNA in human throat swabs. Uses loop-mediated isothermal DNA amplification (LAMP) to target segment of the M. pneumoniae genome

  • Results in less than 1 hour
  • Detects Mycoplasma pneumoniae on day 1 of symptoms
  • Simple sample preparation from throat swabs
  • High sensitivity and specificity
  • 0.0% Invalid Rate
  • Quicker initiation of appropriate patient care
  • Expanded molecular menu
  • No capital expense or service contract required
  • Small footprint, easily adaptable to any laboratory

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance CE Marked, FDA Cleared
Content And Storage 2°C to 27°C
Description FDA Cleared
CE Marker Yes
Clia Complexity Moderate
Sample Type Throat Swab
Sensitivity 96%
Quantity 50 Tests
Specificity 0.977
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.