Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alexa Fluor™ 488 Mouse Anti-Human Chromogranin A, BD Pharmingen™

Catalog No. BDB570152 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB570152 25 Tests
BDB570071 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB570152 Supplier BD Biosciences Supplier No. 570152
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Chromogranin A Monoclonal antibody specifically detects Chromogranin A in Human samples. It is validated for Intracellular staining (flow cytometry)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chromogranin A
Applications Flow Cytometry
Classification Monoclonal
Clone S21-537
Conjugate Alexa Fluor 488
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias CGA; CgA; CHGA; Chromogranin-A; CMGA; SP-I
Host Species Mouse
Immunogen Human Chromogranin A Recombinant Protein
Purification Method Affinity Chromatography
Quantity 25 Tests
Regulatory Status RUO
Gene ID (Entrez) 1113
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.