Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alexa Fluor™ 488 Mouse Anti-Human IL-13 Receptor α2 (CD213a2), BD Pharmingen™

Catalog No. BDB570112 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB570112 25 Tests
BDB570031 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB570112 Supplier BD Biosciences Supplier No. 570112
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-13 Receptor α2 (CD213a2) Monoclonal antibody specifically detects IL-13 Receptor α2 (CD213a2) in Human samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-13 Receptor α2 (CD213a2)
Applications Flow Cytometry
Classification Monoclonal
Clone 47 (also known as 47; mAb47; 47 mAb)
Conjugate Alexa Fluor 488
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias CD213A2; IL-13 receptor subunit alpha-2; IL-13R; IL-13Rα2; cancer/testis antigen 19
Host Species Mouse
Immunogen Human rhIL13Ra2hFc
Purification Method Affinity Chromatography
Quantity 25 Tests
Regulatory Status RUO
Gene ID (Entrez) 3598
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.