Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Alexa Fluor™ 488 Reactive Dye Decapack, for microarrays

Catalog No. a32750
Encompass_Preferred
Click to view available options
Quantity:
1 Set

Matches the imaging channels of most popular microarray scanners

The Alexa Fluor™ reactive dye decapack is specially packaged succinimidyl ester dye for labeling of DNA or RNA samples that have been modified by incorporation of aminoallyl-dUTP (Cat. No. A21664) or aminohexylacrylamido-dUTP (Cat. No. A32761). Every packaging lot is rigorously tested for the ability to label aminoallyl-modified DNA to high and consistent levels.

Array CGH, DNA and RNA Purification and Analysis, DNA Labeling, Expression Array Labeling, Gene Expression Analysis and Genotyping, Genotyping and Genomic Profiling, Microarray Analysis, Nucleic Acid Labeling and Oligo Synthesis

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Product Type Reactive Dye
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Label or Dye Alexa Fluor Dye
Labeling Method Indirect Labeling
Shipping Condition Room Temperature
Product Line Alexa Fluor™
Quantity 1 Set

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.