Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Probes™ Alexa Fluor™ 488 TFP ester

Catalog No. p-7075607
Encompass_Preferred
Click to view available options
Quantity:
25 mg
3 x 100 μg

Alexa Fluor™ dyes are reactive molecules that can be used to add a fluorescent label to the primary amines (R-NH2) of proteins, amine-modified oligonucleotides, and other amine-containing molecules.

Alexa Fluor™ dyes are reactive molecules that can be used to add a fluorescent label to the primary amines (R-NH2) of proteins, amine-modified oligonucleotides, and other amine-containing molecules. The resulting Alexa Fluor™ conjugates exhibit brighter fluorescence and greater photostability than the conjugates of other spectrally similar fluorophores. Alexa Fluor™ 488 TFP ester produces a conjugate with excitation/emission of 495/515 nm that is spectrally similar to fluorescein (FITC) and Cy2 conjugates.

Alexa Fluor™ dyes are available with fluorescence emissions that span the visible and near-infrared spectrum (see The Alexa Fluor™ Dye Series—Note 1.1 in The Molecular Probes™ Handbook) and provide the unique combination of water solubility and pH insensitivity between pH 4 and 10 for compatibility in diverse biological environments.

TFP (tetrafluorophenyl) esters are an improvement over the succinimidyl ester (SE or NHS-ester) chemistry typically used to attach fluorophores or haptens to the primary amines of biomolecules. Both reactive chemistries produce the same strong amide bond between the dye or hapten and the compound of interest, but TFP esters are less susceptible to spontaneous hydrolysis during conjugation reactions. Alexa Fluor™ TFP esters are stable for several hours at the basic pH typically used for reactions–far outlasting succinimidyl esters.

Typical Conjugation Reaction
You can conjugate amine-reactive reagents with virtually any protein or peptide; the provided protocol is optimized for IgG antibodies. You may scale the reaction for any amount of protein, but the concentration of the protein should be at least 2 mg/mL for optimal results. We recommend trying three different degrees of labeling, using three different molar ratios of the reactive reagent to protein.

The Alexa Fluor™ TFP ester is typically dissolved in high-quality, anhydrous dimethylsulfoxide (DMSO) (D12345) , and the reaction is carried out in 0.1–0.2 M sodium bicarbonate buffer, pH 8.3, at room temperature for 1 hour. Because the pKa of the terminal amine is lower than that of the lysine epsilon-amino group, you may achieve more selective labeling of the amine terminus using a buffer closer to neutral pH.

Conjugate Purification
Labeled antibodies are typically separated from free Alexa Fluor™ dye using a gel filtration column, such as Sephadex™ G-25, BioGel™ P-30, or equivalent. For much larger or smaller proteins, select a gel filtration media with an appropriate molecular weight cut-off or purify by dialysis.

Learn More About Protein and Antibody Labeling
We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See Antibody Labeling from A to Z or use our Labeling Chemistry Selection Tool for other choices. To learn more about our labeling kits, read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in The Molecular Probes™ Handbook.

We’ll Make a Custom Conjugate for You
If you can’t find what you’re looking for in our online catalog, we’ll prepare a custom antibody or protein conjugate for you. Our custom conjugation service is efficient and confidential, and we are ISO 13485:2000 certified.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 3 x 100 μg
Product Type Dye
Content And Storage 3 tubes
Store dessicated at -20°C, upon reciept
Shipping Condition Room Temperature
Product Line Alexa Fluor™
Emission 515 nm
Excitation 495 nm
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.