Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Alexa Fluor™ 488 Tyramide SuperBoost™ Kit, goat anti-mouse IgG

Catalog No. p-7083197
Encompass_Preferred
Click to view available options
Quantity:
Sufficient for 150 Slides
Sufficient for 50 Slides

Features poly-HRP-conjugated goat anti-mouse IgG secondary antibody. Alexa Fluor™ 488 Tyramide SuperBoost™ Kit combine brightness of AlexaFluor dyes with high signal amplification of a poly-HRP-mediated tyramide labeling reaction to produce se

SuperBoost™ tyramide signal amplification is most sensitive method for detection of low abundant targets in multiplexable fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC ), and in situ hybridization (ISH).

Features:

  • High sensitivity for detection of low-level or hard-to-detect targets by fluorescent imaging
  • Simple protocol and detection using standard filters
  • Suitable for high-resolution multiplex images, co-label with DAPI, secondary antibodies, and other SuperBoost kits
  • Requires 10-100 times less primary antibody then standard ICC/IHC/ISH experiments

Benefits:

  • Enhancement of signal using Alexa Fluor tyramides: Utilize Alexa Fluor tyramides, which react with HRP to ultimately deposit bright and photostable Alexa Fluor dye on surrounding proteins and other similar molecules
  • Poly-HRP enhancement: Several HRP enzymes are conjugated with short polymers, enhancing signal by several fold over regular HRP systems
  • Reaction stop solution: Include an HRP stop solution to halt the HRP reaction
  • Reduction of background: Include blockers for elimination or reduction of endogenous peroxidase and fluorescent background signals

Shelf Life:

  • Stable for 6 months after receiving
TRUSTED_SUSTAINABILITY

Specifications

Conjugate Alexa Fluor 488
Quantity Sufficient for 150 Slides
Content And Storage 1 kit sufficient for 150 microscope slides (18 x 18 mm), containing: Blocking buffer (1X), 22.5 mL
  • Poly-HRP-conjugated goat anti-mouse secondary antibody (1X), 22.5 mL
  • Alexa Fluor tyramide reagent
  • Hydrogen perox
  • Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
    Product Line SuperBoost™
    Product Type Tyramide SuperBoost Kit

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.