Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alexa Fluor™ 647 Biosimilar Anti-Human CD20, BD Pharmingen™
Biosimilar Monoclonal Antibody
Supplier: BD Biosciences 570531
Description
CD20 Monoclonal antibody specifically detects CD20 in Human samples. It is validated for Flow cytometry
Specifications
CD20 | |
Monoclonal | |
Alexa Fluor 647 | |
B1; Bp35; LEU-16; MS4A1 | |
Human CD20 | |
25 Tests | |
931 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Flow Cytometry | |
Obinutuzumab297.rMAb (also known as Obinutuzumab N297A Biosimilar.rMAb) | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction