Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alexa Fluor™ 647 Biosimilar Anti-Human CD52, BD Pharmingen™
Biosimilar Monoclonal Antibody
$171.20 - $379.00
Specifications
| Antigen | CD52 |
|---|---|
| Clone | Alemtuzumab297.rMAb (also known as Alemtuzumab N297A Biosimilar.rMAb) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Alexa Fluor 647 |
Description
CD52 Monoclonal antibody specifically detects CD52 in Human samples. It is validated for Flow cytometrySpecifications
| CD52 | |
| Flow Cytometry | |
| Alexa Fluor 647 | |
| Mouse | |
| Human | |
| CAMPATH-1 antigen; HE5 | |
| Human CD52 | |
| Affinity Chromatography |
| Alemtuzumab297.rMAb (also known as Alemtuzumab N297A Biosimilar.rMAb) | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| 1043 | |
| IgG1 κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title