Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alexa Fluor™ 647 Mouse Anti-Human CD34, BD Pharmingen™

Catalog No. BDB568771 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB568771 100 Tests
BDB568772 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB568771 Supplier BD Biosciences Supplier No. 568771
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD34 Monoclonal antibody specifically detects CD34 in Human samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD34
Applications Flow Cytometry
Classification Monoclonal
Clone QBEND/10.rMAb (also known as QB-END-10, QBEND10, QBEND/10, QBEnd10)
Conjugate Alexa Fluor 647
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias CD34; CD34 antigen
Host Species Mouse
Immunogen Human Placental Vesicles
Purification Method Affinity Chromatography
Quantity 100 Tests
Regulatory Status RUO
Gene ID (Entrez) 947
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG1 λ2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.