Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alexa Fluor™ 647 Mouse Anti-Human Chromogranin A, BD Pharmingen™
Mouse Monoclonal Antibody
$154.10 - $379.00
Specifications
| Antigen | Chromogranin A |
|---|---|
| Clone | S21-537 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Alexa Fluor 647 |
Description
Chromogranin A Monoclonal antibody specifically detects Chromogranin A in Human samples. It is validated for Intracellular staining (flow cytometry)Specifications
| Chromogranin A | |
| Flow Cytometry | |
| Alexa Fluor 647 | |
| Mouse | |
| Human | |
| CGA; CgA; CHGA; Chromogranin-A; CMGA; SP-I | |
| Human Chromogranin A Recombinant Protein | |
| Affinity Chromatography |
| S21-537 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| 1113 | |
| IgG1 κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title