Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alexa Fluor™ 647 Mouse Anti-Human Integrin αVβ6, BD Pharmingen™

Catalog No. BDB568397 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB568397 25 μg
BDB568396 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB568397 Supplier BD Biosciences Supplier No. 568397
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin αVβ6 Monoclonal antibody specifically detects Integrin αVβ6 in Human|Mouse samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin αVβ6
Applications Flow Cytometry
Classification Monoclonal
Clone 10D5
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 647
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias αVβ6; avb6; CD51/β6; ITGAV/ITGB6; Integrin alpha v beta 6
Host Species Mouse
Immunogen 129/C57BL/6 β6 -/- Mice were mmunized with Human αvβ6 or Mouse Keratinocytes
Purification Method Affinity Chromatography
Quantity 25 μg
Regulatory Status RUO
Gene ID (Entrez) 3685, 3694
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.