Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alexa Fluor™ 647 Mouse Anti-Mouse TCR Vγ7, BD Pharmingen™
Mouse Monoclonal Antibody
$84.00 - $174.00
Specifications
Antigen | TCR Vγ7 |
---|---|
Clone | F2.67 (also known as F4.67) |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
TCR Vγ7 Monoclonal antibody specifically detects TCR Vγ7 in Mouse samples. It is validated for Flow cytometrySpecifications
TCR Vγ7 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
21641 | |
IgG2a κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
F2.67 (also known as F4.67) | |
Flow Cytometry | |
Alexa Fluor 647 | |
Mouse | |
Mouse | |
T Cell Receptor Vγ7; TCR Vg7; TCR Vgamma7; Tcrg-V7; Trgv7 | |
Mouse γδ T Cell Hybridoma Cells | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title