Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alexa Fluor™ 647 Rabbit Anti-Human LDH, BD Pharmingen™

Catalog No. BDB570529 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB570529 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB570529 Supplier BD Biosciences Supplier No. 570529
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

LDH Monoclonal antibody specifically detects LDH in Human samples. It is validated for Intracellular staining (flow cytometry)
TRUSTED_SUSTAINABILITY

Specifications

Antigen LDH
Applications Flow Cytometry
Classification Monoclonal
Clone EP1563Y
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 647
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias LDH heart subunit; LDH muscle subunit; LDH testis subunit; LDH-H; LDH-M; LDH-X; LDHA; LDHB; LDHC
Host Species Rabbit
Immunogen Not Reported
Purification Method Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Gene ID (Entrez) 3939, 3945, 3948
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.