Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alexa Fluor™ 647 Rat Anti-β-galactosidase, BD Pharmingen™
Rat Monoclonal Antibody
Supplier: BD Biosciences 570541
Description
β-galactosidase Monoclonal antibody specifically detects β-galactosidase in Bacteria samples. It is validated for Intracellular staining (flow cytometry)
Specifications
β-galactosidase | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Chromatography | |
RUO | |
Bacteria | |
Liquid |
Flow Cytometry | |
13R4.rMAb (also known as 13R4) | |
Alexa Fluor 647 | |
beta-gal; beta-galactosidase; lacZ; lactase; β-gal; β-galactosidase | |
E. coli β-galactosidase | |
25 μg | |
945006 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction