Learn More
Amberlite™ IRN-150, ion exchange resin
Description
Amberlite IRN-150 is used in water treatment and purification in primary side chemical volume control systems (CVS) in PWR nuclear power generating facilities. It is also used in reactor water clean-up and radio active waste systems. It is recommended in any non-regenerable mixed bed application and can also be used for a variety of electroplating industry waste regeneration applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Beads |
| Chemical Name or Material | Amberlite IRN-150 |
| Odor | Odorless |
| Molecular Formula | Styrene-DVB |
| MDL Number | MFCD00145822 |
| Color | Amber |
| Quantity | 500 g |
| Solubility Information | Insoluble in water,acids and bases. |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.