Learn More
Formaldehyde, Quant™ Test Strips
Catalog No. aa23277qa
Click to view available options
Quantity:
1 Kit
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
Range Measures 0 to 100ppm HCHO in aqueous solution
Type Formaldehyde test strip
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.