Learn More
Thermo Scientific Chemicals Silica gel, wide pore, 150 angstroms
Catalog No. p-4765045
Click to view available options
Quantity:
50 g
250 g
1 kg
5 kg
Catalog No. AA4272618
Supplier: Thermo Scientific Chemicals 04272618
Only null left
Add to Cart
Silica gel, wide pore is mainly used for the dehydrated purification of the industrial gases, the clearance of the organic acids and high polymers in the insulative oil, the purification and separation of the medicines. It is also used in column chromatography without applied pressure (gravity pulled).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Particle Size -100/200 Mesh
Pore Size 150 Å
Packaging Bottle
pH 6.5 to 7.5
Quantity 50 g
Surface Area (Metric) 350 to 400 m2/g
Type Silica Gel
Material Silica
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.