Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ALG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543317
Description
Peptide sequence: VKHEENGLVF EDSEELAAQL QMLFSNFPDP AGKLNQFRKN LRESQQLRWD Sequence homology: Cow: 86%; Dog: 93%; Guinea Pig: 92%; Human: 100%; Mouse: 86%; Rabbit: 86%; Rat: 92%.
The enzyme encoded by this gene catalyzes the first mannosylation step in the biosynthesis of lipid-linked oligosaccharides. This gene is mutated in congenital disorder of glycosylation type Ik.
Specifications
| ALG1 | |
| Polyclonal | |
| Unconjugated | |
| ALG1 | |
| Alg1; alg-1; ALG1 chitobiosyldiphosphodolichol beta-mannosyltransferase; ALG1, chitobiosyldiphosphodolichol beta-mannosyltransferase; Argonaute; Argonaute-like; asparagine-linked glycosylation 1 (beta-1,4-mannosyltransferase); asparagine-linked glycosylation 1 homolog (S. cerevisiae, beta-1,4-mannosyltransferase); asparagine-linked glycosylation 1 homolog (yeast, beta-1,4-mannosyltransferase); asparagine-linked glycosylation 1, beta-1,4-mannosyltransferase homolog; asparagine-linked glycosylation protein 1 homolog; beta-1,4 mannosyltransferase; beta-1,4-mannosyltransferase; CDG1K; CELE_F48F7.1; chitobiosyldiphosphodolichol beta-mannosyltransferase; GDP-Man:GlcNAc2-PP-dolichol mannosyltransferase; GDP-mannose-dolichol diphosphochitobiose mannosyltransferase; HMAT1; hMat-1; HMT1; HMT-1; Mannosyltransferase-1; Mat-1; MT-1; Protein argonaute; protein-coding; PSEC0061; UNQ861/PRO1870 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 56052 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BT22 | |
| ALG1 | |
| Synthetic peptide directed towards the C-terminal of human ALG1 (aa 401-450). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction