Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563853
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563853 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563853 Supplier Invitrogen™ Supplier No. PA563853
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NWHIRAVTVY DKPASFFKET PLDLQHRLFM KLGSMHSPFR ARSEPEDPVT ERSAFTERDA GSGLVTRLRE RP Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 72%.

The enzyme encoded by this gene catalyzes the first mannosylation step in the biosynthesis of lipid-linked oligosaccharides. This gene is mutated in congenital disorder of glycosylation type Ik.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALG1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ALG1
Gene Accession No. Q9BT22
Gene Alias Alg1; alg-1; ALG1 chitobiosyldiphosphodolichol beta-mannosyltransferase; ALG1, chitobiosyldiphosphodolichol beta-mannosyltransferase; Argonaute; Argonaute-like; asparagine-linked glycosylation 1 (beta-1,4-mannosyltransferase); asparagine-linked glycosylation 1 homolog (S. cerevisiae, beta-1,4-mannosyltransferase); asparagine-linked glycosylation 1 homolog (yeast, beta-1,4-mannosyltransferase); asparagine-linked glycosylation 1, beta-1,4-mannosyltransferase homolog; asparagine-linked glycosylation protein 1 homolog; beta-1,4 mannosyltransferase; beta-1,4-mannosyltransferase; CDG1K; CELE_F48F7.1; chitobiosyldiphosphodolichol beta-mannosyltransferase; GDP-Man:GlcNAc2-PP-dolichol mannosyltransferase; GDP-mannose-dolichol diphosphochitobiose mannosyltransferase; HMAT1; hMat-1; HMT1; HMT-1; Mannosyltransferase-1; Mat-1; MT-1; Protein argonaute; protein-coding; PSEC0061; UNQ861/PRO1870
Gene Symbols ALG1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ALG1. Recombinant protein control fragment (Product #RP-101918).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56052
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.