Learn More
Invitrogen™ ALG1 Polyclonal Antibody

Description
Immunogen sequence: YDKPASFFKE TPLDLQHRLF MKLGSMHSPF RARSEPEDPV TERSAFTERD AGSGLVTRLR ERPALLVSST SWTEDEDFSI LLAALEKFEQ LTLDGHNLPS LVCVITGKGP LREYYSRLIH QKHFQHIQVC TPWLEAEDYP LLLGSADLGV CLHTSSSGLD LPMKVVDMFG CCLPVCAVNF KCLHELVKHE ENGLVFEDSE ELAAQLQMLF SNFPDPAGKL NQFRKNLRES QQLRWDESWV QTVLPLVMDT; Positive Samples: HeLa, HepG2, Mouse liver, Mouse kidney, Mouse ovary, Rat liver; Cellular Location: Endoplasmic reticulum membrane, Single-pass type II membrane protein.
Specifications
Specifications
| Antigen | ALG1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.7 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ALG1 |
| Gene Accession No. | Q921Q3, Q9BT22 |
| Gene Alias | Alg1; alg-1; ALG1 chitobiosyldiphosphodolichol beta-mannosyltransferase; ALG1, chitobiosyldiphosphodolichol beta-mannosyltransferase; Argonaute; Argonaute-like; asparagine-linked glycosylation 1 (beta-1,4-mannosyltransferase); asparagine-linked glycosylation 1 homolog (S. cerevisiae, beta-1,4-mannosyltransferase); asparagine-linked glycosylation 1 homolog (yeast, beta-1,4-mannosyltransferase); asparagine-linked glycosylation 1, beta-1,4-mannosyltransferase homolog; asparagine-linked glycosylation protein 1 homolog; beta-1,4 mannosyltransferase; beta-1,4-mannosyltransferase; CDG1K; CELE_F48F7.1; chitobiosyldiphosphodolichol beta-mannosyltransferase; GDP-Man:GlcNAc2-PP-dolichol mannosyltransferase; GDP-mannose-dolichol diphosphochitobiose mannosyltransferase; HMAT1; hMat-1; HMT1; HMT-1; Mannosyltransferase-1; Mat-1; MT-1; Protein argonaute; protein-coding; PSEC0061; UNQ861/PRO1870 |
| Gene Symbols | ALG1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.